ACVR1 (ALK2) Recombinant Human Protein DFS Item
ACVR1 (ALK2) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4877
 View more versions of this product
Catalog No. PV4877
Description
Description
Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors
Specifications
Specifications
NP_001096.1
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
69.1kDa
90% by SDS-PAGE
–80°C
1 Each
ACVR1 (ALK2)
ACVR1 (ALK2) was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 12 MnCl2, 1.2mM EGTA, 1mM Na3VO4, 5XmM β-glycerophosphate, 2mM DTT, 0.02% Triton™ X-100, 200μM ATP, 500μg/mL dephosphorylated casein substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
STKR
ACVR1 (ALK2) Recombinant Human Protein
Human
0.4mg/mL
90
ACVR1 (ALK2) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
ACVR1 (ALK2) Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 145-509
580nmoles of phosphate transferred to dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.417μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.