Learn More
ACVR1 (ALK2) [R206H] Recombinant Human Protein
ACVR1 (ALK2) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins which encodes activin A type I receptor. Recombinant human ACVR1 (R206H) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Supplier: Thermo Fisher Scientific PV6233
Description
ACVR1 signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 are associated with fibrodysplasia ossificans progressiva (FOP) and the mutations are located in or adjacent to the GS regulatory region or active site of the kinase. The mutations in ACVR1 were predicted by protein structure homology modeling to activate the ACVR1 protein and enhance receptor signaling. Dysregulated ACVR1 kinase activity plays a role in the pathogenesis of FOP.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
67.6kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 147-end | |
Recombinant |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol | |
100μg | |
ACVR1, FOP, SKR1, TSRI, ACTRI, ACVR1A, ACVRLK2 | |
Human | |
GST Tag |