Please login to your online account to display your discounted pricing
Thermo Scientific™ ActivX™ Fluorophosphonate Probes
Tagged phosphonate probes to assay serine hydrolase active sites using fluorescence, biotin-affinity purification or detection, or mass spectrometry.
$571.71 - $763.02
Specifications
Sufficient For 100 labeling reactions or 10 pull-down assays
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    
PI88316
thermo scientific™
88316*
Azido-fluorophosphonate, MW 350 Each for $763.02
PI88317
thermo scientific™
88317
Desthiobiotin-fluorophosphonate, MW 464 Each for $571.71
PI88318
thermo scientific™
88318
TAMRA-fluorophosphonate, MW 680 Each for $571.71
Description & Specifications
Specifications
Sufficient For 100 labeling reactions or 10 pull-down assays
Thermo Scientific™ ActivX Fluorophosphonate (FP) Probes allow covalent labeling of the serine hydrolase active site with affinity, fluorescent or reactive tags to facilitate selective enrichment, detection or multiplex labeling.
ActivX FP Probes feature a reactive fluorophosphonate group that specifically and covalently labels the active-site serine of enzymatically active serine hydrolases. These probes are available with a desthiobiotin (biotin analog) tag for selective enrichment, TAMRA fluorophore for detection or a reactive- azido group (Staudinger reagents) that facilitates multiplex labeling when used with phosphine- or alkyne-derivatized tags. These probes can be used to assess activity or screen small molecule inhibitors against enzymes derived from cell lysates, subcellular fractions, tissues and recombinant proteins.
Highlights:
  • Specific – labels the reactive site of active serine hydrolases
  • Compatible – tags available for capture, detection and Staudinger conjugation
  • Flexible – use for in vitro or intracellular enzyme labeling
Recommended for:
Determine serine hydrolase enzyme activity in cells and lysates; Mapping the active-site serine of functionally diverse serine hydrolase family members (e.g. proteases, lipases, esterases); Screen for small molecule binding affinities and active-site inhibition; Profile serine hydrolases using fluorescent, Western blot or mass spectrometry workflows