missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Human Activin A Recombinant Protein
Activin A Recombinant Human Protein
Supplier: Gibco™ PHC9561
Description
Human Activin A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Recombinant Human Activin A is a bioactive protein intended for use in cell culture applications. Activin A is involved in multiple biological processes including inflammation, neural development, and haematopoiesis.High purity recombinant bioactive protein
Recombinant Human Activin A is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High biological activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Activin A specific activity is determined by the dose dependent proliferation of mouse MPC-11 cells.
Proven compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Specifications
Cell Culture | |
Gel-Purified | |
Activin A | |
Human | |
Carrier-Free | |
Inhibition of Proliferation of MPC11 cells | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation | |
95 % |
25 kDa | |
100μg | |
116 aa | |
Store in freezer (-5 to -30°C). | |
0.5 ng/ml | |
< 1 EU/ml | |
Mammalian | |
Recombinant, Ligand | |
Activins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.