missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Activated Charcoal Sorbent, EMD-Millipore
Supplier: Technidata 1096310500
Specifications
Activated Charcoal Sorbent | |
Glass | |
C | |
[C] | |
12.011 | |
CHEBI:27594 |
7440-44-0 | |
500 g | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01 |
Safety and Handling
Recommended Storage : Store without restrictions