missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Activated Charcoal Sorbent, EMD-Millipore

Manufacturer:  MilliporeSigma 1096310100

 View more versions of this product

Catalog No. M1096310100



Specifications

Specifications

Activated Charcoal Sorbent
150 to 440kg/m3
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12.01
100g
7440-44-0
C
[C]
12.011
CHEBI:27594
Glass

Chemical Identifiers

7440-44-0
12.011
5462310
carbon
C
OKTJSMMVPCPJKN-UHFFFAOYSA-N
CHEBI:27594
[C]
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

Documents