missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Activated Charcoal Sorbent, EMD-Millipore
$336.34 - $1,663.89
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.011 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1096310100
|
Emd Millipore Corporation
1096310100 |
100g | Glass |
Each for $336.34
|
|
M1096310500
|
Emd Millipore Corporation
1096310500 |
500g | Glass |
Each for $1,663.89
|
|
Chemical Identifiers
7440-44-0 | |
12.011 | |
5462310 | |
carbon |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
C | |
[C] | |
12.011 | |
CHEBI:27594 | |
Glass | |
Activated Charcoal Sorbent |
150 to 440kg/m3 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01 | |
100g |
Safety and Handling
Recommended Storage : Store without restrictions