missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Activated charcoal, NORIT(R) SA 2, decolorizing, Acros Organics

$126.85 - $419.22

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym acticarbone
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC171530010 SDS View Documents Fisher Scientific Acros
171530010
1kg Plastic drum Each for $126.85
 
AC171530050 SDS View Documents Thermo Fisher Scientific
AC171530050
5kg Plastic drum Each for $419.22
 
Description

Description

Filter aid
Specifications

Chemical Identifiers

7440-44-0
12
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
acticarbone
CHEBI:27594
[C]

Specifications

10% max.
Black
1kg
C
15, 1807
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
[C]
12
CHEBI:27594
Powder
Plastic drum
3652.0°C
7440-44-0
MFCD00133992
acticarbone
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12
Activated charcoal,NORIT&asterisk; SA 2
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3