missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Activated charcoal, NORIT(R) GAC 1240, 12-40 mesh, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 395712500

 View more versions of this product

Catalog No. AC395712500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
Specifications

Specifications

Activated charcoal,NORIT* GAC 124
3652.0°C
2% max. (as packed)
Glass bottle
MFCD00133992
15, 1807
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
[C]
12
CHEBI:27594
Granular Solid
7440-44-0
Black
12 mesh (min. 5%), 40 mesh (max. 4% max.)
C
250 g
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12

Chemical Identifiers

7440-44-0
12
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only