Learn More
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807
Supplier: Thermo Scientific Chemicals 134342500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidSpecifications
Activated charcoal,NORIT* A SUPRA | |
7440-44-0 | |
Black | |
Glass bottle | |
MFCD00133992 | |
15, 1807 | |
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5% | |
[C] | |
12 | |
CHEBI:27594 | |
Powder |
Passes Test | |
3652.0°C | |
15% max. | |
C | |
250 g | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12 |
Chemical Identifiers
7440-44-0 | |
12 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Safety and Handling
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : TSCA
RUO – Research Use Only