Please login to your online account to display your discounted pricing

Activated charcoal, iodinated, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Activated charcoal
Assay 5%, exact strength on the certificate of analysis
Molecular Formula C
Formula Weight 12
MDL Number MFCD00133992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389470010

  SDS
thermo fisher scientific
AC389470010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC389472500

  SDS
thermo fisher scientific
AC389472500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Thin Rods
Formula Weight 12
Quantity 1kg
Color Black
Merck Index 14, 1807
Assay 5%, exact strength on the certificate of analysis
MDL Number MFCD00133992
Chemical Name or Material Activated charcoal
Packaging Glass bottle
Molecular Formula C
CAS 7440-44-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Health Hazard 2 GHS P Statement
Flammable solid.

Filter aid

Safety and Handling

Warning

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement
Flammable solid.