Please login to your online account to display your discounted pricing

Activated charcoal, DARCO™ G-60, decolorizing, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Activated charcoal, DARCO™ G-60
CAS 7440-44-0
Molecular Formula C
Formula Weight 12
MDL Number MFCD00133992
Synonym Carbon
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC403980050

  SDS
thermo fisher scientific
AC403980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 12
Quantity 5g
Color Black
TSCA TSCA
RTECS Number FF5250100
EINECS Number 231-153-3
Loss on Drying 12% max.
Packaging Glass bottle
Molecular Formula C
Water Soluble Substances 0.5% max.
Synonym Carbon
MDL Number MFCD00133992
CAS 7440-44-0
Chemical Name or Material Activated charcoal, DARCO™ G-60
Iron (Fe) 200ppm max.
Additional Information methylene blue adsorption (g/100g):> 15

Filter aid