Please login to your online account to display your discounted pricing

Activated charcoal, DARCO™ G-60, decolorizing, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC403980050

 View more versions of this product

Catalog No. AC403980050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 12
Quantity 5g
Color Black
TSCA TSCA
RTECS Number FF5250100
EINECS Number 231-153-3
Loss on Drying 12% max.
Packaging Glass bottle
Molecular Formula C
Water Soluble Substances 0.5% max.
Synonym Carbon
MDL Number MFCD00133992
CAS 7440-44-0
Chemical Name or Material Activated charcoal, DARCO™ G-60
Iron (Fe) 200ppm max.
Additional Information methylene blue adsorption (g/100g):> 15

Specifications

Chemical Name or Material Activated charcoal, DARCO™ G-60
CAS 7440-44-0
Molecular Formula C
Formula Weight 12
MDL Number MFCD00133992
Synonym Carbon