missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Activated charcoal, DARCO(R), 4-12 mesh, decolorizing, Thermo Scientific Chemicals
$135.69 - $1,214.64
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC403995000
|
Thermo Scientific Chemicals
403995000 |
500 g | Glass bottle |
N/A
|
|
||||
AC403990030
|
Thermo Scientific Chemicals
403990030 |
3 kg | Plastic drum |
N/A
|
|
||||
AC403990100
|
Thermo Scientific Chemicals
403990100 |
10 kg | Plastic drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
7440-44-0 | |
12 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
Black | |
12% max. | |
Glass bottle | |
MFCD00133992 | |
15, 1807 | |
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5% | |
[C] | |
12 | |
CHEBI:27594 | |
Granules or Chunks |
3652.0°C | |
3.5 to 6 | |
> 4 mesh: < 5%, < 12 mesh: < 5% | |
C | |
500 g | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12 | |
Activated charcoal,DARCO*, DARCO™, 4 to 12 Mesh, Decolorizing |
Safety and Handling
EINECSNumber : 231-153-3
RUO – Research Use Only