Please login to your online account to display your discounted pricing

American Educational Products Action/Reaction Lab


Investigate response time and the nervous system.

Manufacturer: american educational products  6027

 View more versions of this product

Catalog No. S16213

  • / Each
 

Description & Specifications

Specifications

Age 8 to 15 years
For Use With (Application) Used to time a person's reaction to light, sound and touch stimuli
Grade Level 3 to 9
Includes Instruction sheet
Product Type Reaction Timer