Please login to your online account to display your discounted pricing

Actinomycin D (Red Crystals), Fisher BioReagents


Isolated from Streptomyces parvuius

Quantity 10mg
Packaging Poly Micro Tube

Manufacturer: fisher scientific company  FLBP60610

 View more versions of this product

Catalog No. BP606-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point 245°C
Physical Form Powder/Solid
Formula Weight 1255.44
Quantity 10mg
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Molecular Formula C62 H86N12O16
Color Red
Decomposition Information 250°C
Solubility Soluble in Methanol
Merck Index 15, 2797
Assay Percent Range ≥98 %
Purity 99.0% min. (HPLC), 98% min. (TLC)
Chemical Name or Material Actinomycin-D
ChemAlert Storage Symbol Blue
CAS 50-76-0
Packaging Poly Micro Tube
Extinction Coefficient Report
Recommended Storage 4°C

Specifications

Chemical Name or Material Actinomycin-D
CAS 50-76-0
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C62 H86N12O16
Formula Weight 1255.44
Accessories
 

4°C

SDS & Safety Documents