Please login to your online account to display your discounted pricing

Actinomycin D, ∽98%, MP Biomedicals™


Quantity 25mg

Manufacturer: mp biomedicals  0219603525

 View more versions of this product

Catalog No. ICN19603525

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25mg
Chemical Name or Material Actinomycin D
Name Note For TLC analysis
CAS 50-76-0
Purity Grade ∽98%
Physical Form Red-orange to red powder
Molecular Formula C62H86N12O16
Formula Weight 1255.5g/mol

Specifications

Chemical Name or Material Actinomycin D
Name Note For TLC analysis
CAS 50-76-0
Purity Grade ∽98%
Physical Form Red-orange to red powder
Molecular Formula C62H86N12O16
Formula Weight 1255.5g/mol