Please login to your online account to display your discounted pricing

Actinomycin D, ∽98%, MP Biomedicals™


Quantity 5mg

Manufacturer: mp biomedicals  19603505

 View more versions of this product

Catalog No. ICN19603505

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ∽98%
Formula Weight 1255.5
Color Red-orange to red
Quantity 5mg
Chemical Name or Material Actinomycin D
Name Note For TLC analysis
Molecular Formula C62H86N12O16
CAS 50-76-0

Specifications

Chemical Name or Material Actinomycin D
Name Note For TLC analysis
CAS 50-76-0
Physical Form Powder
Molecular Formula C62H86N12O16
Formula Weight 1255.5