missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Actinomycin D, ∽98%, MP Biomedicals™

$129.50 - $129.50

Chemical Identifiers

CAS 50-76-0
Physical Form Powder
Molecular Formula C62H86N12O16
Formula Weight 1255.5
Chemical Name or Material Actinomycin D, For TLC analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19603505 Mp Biomedicals
19603505
5mg Each for $129.50
Specifications Description & Specifications

Chemical Identifiers

50-76-0
C62H86N12O16
Actinomycin D, For TLC analysis
Powder
1255.5

Specifications

Powder
Red-orange to red
C62H86N12O16
Actinomycin D, For TLC analysis
1255.5
5mg
50-76-0
∽98%