missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Actinomycin D, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals J60148LB0

Catalog No. AAJ60148LB0



Description

Description

Actinomycin D is used as an anti-neoplastic antibiotic. It is also used to inhibit RNA polymerase and is a potent inducer of apoptosis. It is also used as a potent antibiotic that binds to the GpC steps of DNA. Inhibits DNA-primed RNA polymerase by complexing with DNA via deoxyguanosine residues. A DNA-intercalator with growth inhibitory activity against certain leukemias and sarcomas. At higher concentrations, DNA polymerase is inhibited. Inhibits cell growth and colony formation in synchronized HeLa cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Actinomycin D is used as an anti-neoplastic antibiotic. It is also used to inhibit RNA polymerase and is a potent inducer of apoptosis. It is also used as a potent antibiotic that binds to the GpC steps of DNA. Inhibits DNA-primed R
Specifications

Specifications

Actinomycin D
Powder
251°C to 253°C
MFCD00005033
605235
Dactinomycin; Actinomycin C1
RJURFGZVJUQBHK-HUZQGMAJSA-N
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
131954673
Orange to Red
Actinomycin D
50-76-0
C62H86N12O16
UN3462
14,2800
It is soluble ino 50 mM in DMSO.
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
1255.438
1255.4
5 mg

Chemical Identifiers

50-76-0
1255.438
RJURFGZVJUQBHK-HUZQGMAJSA-N
131954673
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
C62H86N12O16
MFCD00005033
Dactinomycin; Actinomycin C1
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
Safety and Handling

Safety and Handling

GHS H Statement
H300-H360-H351
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.

P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c

H300-H336-H373

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

EINECSNumber : 200-063-6

RTECSNumber : AU1575000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only