Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1296PABX

 View more versions of this product

Catalog No. MS1296PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Actin, Muscle Specific Ab-6
Classification Monoclonal
Clone MSA06; same as HUC1-1
Concentration 1mg/mL
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Purified actin
Isotype IgG1 kappa
Quantity 200μL
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Bovine
Target Species Canine
Target Species Equine
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rat

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MSA06; same as HUC1-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, Pig, Horse, Cow and Dog. Others not tested.

Epitope: Not determined

Immunogen: Purified actin

Positive Control: Muscle or sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO