missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody

Detect muscle-specific actin in immunohistochemistry experiments with Thermo Scientific™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody.

$260.76 - $823.86

Specifications

Antigen Actin, Muscle Specific Ab-4
Clone HHF35
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS742R7 Thermo Scientific™ Lab Vision™
MS742R7
7mL Each for $392.04
MS742S Thermo Scientific™ Lab Vision™
MS742S
1mL Each for $823.86
MS742S0 Thermo Scientific
MS742S0
100μL Each for $260.76
MS742S1 Thermo Scientific
MS742S1
500μL Each for $582.06
Specifications

Description

Detect muscle-specific actin in immunohistochemistry experiments with Thermo Scientific™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: HHF35

Isotype: IgG1/κ

Species Reactivity: Human, Rabbit and Rat. Others not known.

Epitope: Not determined

Immunogen: SDS extract of human myocardium

Positive Control: Muscle or sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Description & Specifications

Specifications

Actin, Muscle Specific Ab-4
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
HHF35
Monoclonal
Murine
Muscle Biology
SDS extract of human myocardium