missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Actin (muscle) Mouse, Clone: HHF35, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $249.48

Specifications

Antigen Actin (muscle)
Clone HHF35
Host Species Murine
Species Reactivity Bovine, Canine, Guinea Pig, Horse, Human, Mouse, Non-human primate, Pig, Rat
Applications Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245578 Life Technologies
LS14649680
25μg Each for $117.48
50245579 Life Technologies
LS14649682
100μg Each for $249.48
Specifications

Description

The monoclonal antibody HHF35 recognizes muscle actin; specifically the alpha and gamma form found in skeletal cardiac, and smooth muscle. No reactivity to the beta form is observed. The 42kDa protein is a main component of the cytoskeletal structural network. The HHF35 antibody has been shown to recognize actin from smooth muscle cells of the heart, bladder, uterus, gastrointestinal tract, and diaphragm . The antibody stains tumors of muscle origin such as leiomyomas, leiosarcomes and rhabdomyosacromas.

The HHF35 antibody recognizes human, mouse, rat, bovine, pig, guinea pig, sheep, frog, canine and non-human primate actin.
Description & Specifications

Specifications

Actin (muscle)
Murine
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
HHF35
Bovine, Canine, Guinea Pig, Horse, Human, Mouse, Non-human primate, Pig, Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer