Please login to your online account to display your discounted pricing

Actin (muscle) Mouse, Alexa Fluor 488, Clone: HHF35, eBioscience™

Mouse Monoclonal Antibody

$185.60 - $362.50

Specifications

Antigen Actin (muscle)
Clone HHF35
Host Species Mouse
Species Reactivity Bovine, Canine, Guinea Pig, Horse, Human, Mouse, Non-human primate, Pig, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124608

 
life technologies
LS53649680
25μg Each for $185.60

501124609

 
life technologies
LS53649682
100μg Each for $362.50
Description & Specifications

Specifications

Antigen Actin (muscle)
Clone HHF35
Host Species Mouse
Species Reactivity Bovine, Canine, Guinea Pig, Horse, Human, Mouse, Non-human primate, Pig, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody HHF35 recognizes muscle actin; specifically the alpha and gamma form found in skeletal cardiac, and smooth muscle. No reactivity to the beta form is observed. The 42kDa protein is a main component of the cytoskeletal structural network. The HHF35 antibody has been shown to recognize actin from smooth muscle cells of the heart, bladder, uterus, gastrointestinal tract, and diaphragm. The antibody stains tumors of muscle origin such as leiomyomas, leiosarcomes and rhabdomyosacromas.

The HHF35 antibody recognizes human, mouse, rat, bovine, canine and non-human primate actin.