Actin (muscle), Alexa Fluor® 488, Clone: HHF35, eBioscience™
Mouse Monoclonal Antibody
Manufacturer:  Life Technologies LS53649680
 View more versions of this product
Catalog No. 501124608
Description
Description
Description: The monoclonal antibody HHF35 recognizes muscle actin; specifically the alpha and gamma form found in skeletal cardiac, and smooth muscle. No reactivity to the beta form is observed. The 42 kDa protein is a main component of the cytoskeletal structural network. The HHF35 antibody has been shown to recognize actin from smooth muscle cells of the heart, bladder, uterus, gastrointestinal tract, and diaphragm. The antibody stains tumors of muscle origin such as leiomyomas, leiosarcomes and rhabdomyosacromas. The HHF35 antibody recognizes human, mouse, rat, bovine, canine and non-human primate actin. Applications Reported: This HHF35 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This HHF35 antibody has been tested by immunocytochemistry and can be used at less than or equal to 30 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered.
Description: The monoclonal antibody HHF35 recognizes muscle actin; specifically the alpha and gamma form found in skeletal cardiac, and smooth muscle. No reactivity to the beta form is observed. The 42kDa protein is a main component of the cytoskeletal structural network. The HHF35 antibody has been shown to recognize actin from smooth muscle cells of the heart, bladder, uterus, gastrointestinal tract, and diaphragm. The antibody stains tumors of muscle origin such as leiomyomas, leiosarcomes and rhabdomyosacromas.The HHF35 antibody recognizes human, mouse, rat, bovine, canine and non-human primate actin.Applications Reported: This HHF35 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.Applications Tested: This HHF35 antibody has been tested by immunocytochemistry and can be used at less than or equal to 30 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications
Specifications
Actin (muscle)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P62739, P68138, Q3ZC07, P62736, P68032, P68133, P62737, P68033, P68134, P68137, P62738, P68035, P68136
actin, actin alpha 1, actin alpha cardiac, actin alpha cardiac 1, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, alpha, cardiac 1, actin, alpha, vascular smooth muscle, actin, aortic smooth muscle, alpha actin 1, alpha actin 2, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin cardiac, alpha-actin-1, alpha-actin-2, alpha-cardiac actin, alpha-sma, alphac-actin, cell growth-inhibiting gene 46 protein, nemaline myopathy type 3, skeletal alpha actin, smooth muscle alpha-actin
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Bovine, Canine, Equine, Guinea Pig, Human, Murine, Non-human Primate, Porcine, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
HHF35
Alexa Fluor 488
ACTA2
Liquid
ACTA1, Acta2, ACTC1
IgG1, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
100057678, 100062175, 100065454, 100152267, 100154254, 100716933, 100722917, 100731953, 11459, 11464, 11475, 281592, 29275, 29437, 477587, 478250, 488984, 515610, 533219, 58, 59, 70, 733615, 81633
Documents
For Research Use Only.