Please login to your online account to display your discounted pricing

Alfa Aesar™ Acryloyl chloride, 96%, stab. with 400ppm phenothiazine

$56.05 - $319.43

Chemical Identifiers

Chemical Name or Material Acryloyl Chloride
CAS 814-68-6
Name Note stabilized with 400ppm phenothiazine
Molecular Formula C3H3ClO
Linear Formula CH2=CHCOCl
Formula Weight 90.51
MDL Number MFCD00000731
UN Number 3383
Beilstein 635744
Synonym Propenoyl Chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1036318

  SDS
alfa aesar™
L1036318
50g Each for $87.24

AAL1036330

  SDS
alfa aesar™
L1036330
250g Each for $319.43

AAL1036309

  SDS
alfa aesar™
L1036309
10g Each for $56.05
Description & Specifications

Specifications

Chemical Name or Material Acryloyl chloride
Name Note stabilized with 400ppm phenothiazine
CAS 814-68-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
RTECS Number AT7350000
TSCA TSCA
Assay 96%
Molecular Formula C3H3ClO
Linear Formula CH2=CHCOCl
Formula Weight 90.51
MDL Number MFCD00000731
UN Number UN3383
Beilstein 635744
Synonym Propenoyl chloride
Boiling Point 74° to 76°C
Quantity 50g
Flash Point −4°C (24°F)
Odor Acrid

Applications
Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, oxidizing agents, strong bases and polymerizing initiators.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

−30° to −10°C