missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acryloyl chloride, 96%, stab. with 400ppm phenothiazine

Manufacturer:  Alfa Aesar™ L1036309

 View more versions of this product

Catalog No. AAL1036309



Description

Description

Applications
Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, oxidizing agents, strong bases and polymerizing initiators.
Specifications

Specifications

Acryloyl Chloride
Acrid
10g
814-68-6
CH2=CHCOCl
3383
acryloyl chloride, acrylyl chloride, 2-propenoyl chloride, acrylic acid chloride, propenoyl chloride, acryloylchloride, chlorid kyseliny akrylove, unii-8k23o56tg5, chlorid kyseliny akrylove czech, acrylylchloride
C=CC(=O)Cl
90.506
90.51
−4°C (24°F)
74°C to 76°C
stabilized with 400ppm phenothiazine
C3H3ClO
MFCD00000731
635744
HFBMWMNUJJDEQZ-UHFFFAOYSA-N
prop-2-enoyl chloride
13140
96%

Chemical Identifiers

814-68-6
90.506
HFBMWMNUJJDEQZ-UHFFFAOYSA-N
13140
C=CC(=O)Cl
C3H3ClO
MFCD00000731
acryloyl chloride, acrylyl chloride, 2-propenoyl chloride, acrylic acid chloride, propenoyl chloride, acryloylchloride, chlorid kyseliny akrylove, unii-8k23o56tg5, chlorid kyseliny akrylove czech, acrylylchloride
prop-2-enoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

RTECSNumber : AT7350000

TSCA : TSCA

Recommended Storage : −30° to −10°C

Documents