missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Acrylic Glove Box Holder

Clean line storage solutions perfect for laboratory organization. Fisherbrand Acrylic Single Glove Box Holder is made of clear durable acrylic.

Manufacturer:  Fisherbrand™ FH03

 View more versions of this product

Catalog No. 03391575



Description

Description

  • Heat-bent thick walls simplify construction and limit accidental breakage
  • Triple and quad holders have a unique design that allows glove boxes to be loaded from either the top or side
  • Can be mounted vertically or horizontally
  • Fits standard glove boxes accommodating depths up to 3.75 in.
  • Holders offer versatility while providing space-saving storage
  • All mounting accessories included
Specifications

Specifications

Clear
3 glove boxes
Wall
Glove box holder
40.8cm
26.28cm
10.38cm
Holding glove boxes
Mounting accessories
1
16.06 in.
10.35 in.
4.09 in.
Acrylic
Documents