United Scientific Supplies Acrylic Demonstration Lenses Sets
Set of six lenses of a variety of types.
Manufacturer:  United Scientific Supplies LSTA50
 View more versions of this product
Catalog No. S42921
Description
Description
  • Each single-diameter set includes a double convex, plano-convex, concavo-convex, double concave, plano-concave and convexo-concave lens
  • Three diameters available
  • Includes an activity guide
Order Info
Comes in a reusable cardboard storage box.
Specifications
Specifications
2 in.
Teaching and demonstrating the basic principles of optics
6 Pack
50mm
6 Lenses, one each of double convex, plano convex, concavo convex, double concave, plano concave, and convexo concave lens, activity guide and a reusable cardboard storage box
Lens set