Please login to your online account to display your discounted pricing

Alfa Aesar™ Acrylic acid, tech. 90%, stab.

Chemical Identifiers

Chemical Name or Material Acrylic acid
CAS 79-10-7
Name Note Stabilized
Purity Grade Technical Grade
Molecular Formula C3H4O2
Linear Formula H2C=CHCO2H
Formula Weight 72.06
MDL Number MFCD00004367
UN Number UN2218
Beilstein 635743
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45779AP

  SDS
alfa aesar™
45779AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA45779K4

  SDS
alfa aesar™
45779K4
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acrylic acid
Name Note Stabilized
CAS 79-10-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACRYLIC ACID, STABILIZED
RTECS Number AS4375000
TSCA TSCA
Purity Grade Technical Grade
Physical Form Liquid
Assay 90%
Molecular Formula C3H4O2
Linear Formula H2C=CHCO2H
Formula Weight 72.06
MDL Number MFCD00004367
Refractive Index 1.4202
UN Number UN2218
Beilstein 635743
Merck Index 14,130
Synonym Propenoic acid
Density 1.051g/mL
Melting Point ≈13°C
Boiling Point 138° to 139°C
Quantity 500mL
Flash Point 54°C (129°F)
Odor Pungent
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.