missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acrylic acid, tech. 90%, stab.

Manufacturer:  Alfa Aesar™ 45779K4

 View more versions of this product

Catalog No. AA45779K4



Specifications

Specifications

Acrylic acid
54°C (129°F)
1.4202
~13°C
Stabilized
C3H4O2
MFCD00004367
635743
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
C=CC(=O)O
72.063
CHEBI:18308
Liquid
Technical
1.051g/mL
Pungent
138°C to 139°C
2.5L
79-10-7
H2C=CHCO2H
UN2218
14,130
NIXOWILDQLNWCW-UHFFFAOYSA-N
prop-2-enoic acid
6581
72.06
90%

Chemical Identifiers

79-10-7
72.063
NIXOWILDQLNWCW-UHFFFAOYSA-N
6581
prop-2-enoic acid
C3H4O2
MFCD00004367
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
CHEBI:18308
C=CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACRYLIC ACID, STABILIZED

RTECSNumber : AS4375000

TSCA : TSCA

Documents