missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acrylic acid, 98%, extra pure, stabilized, ACROS Organics™

Manufacturer:  Acros Organics AC164250100

 View more versions of this product

Catalog No. AC164250100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acrylic acid
48°C
Plastic drum
1.05
0.1% max. (K.F.)
Undesignated
10L
79-10-7,150-76-5
98%
H2C=CHCO2H
02, 397
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
C=CC(=O)O
72.063
CHEBI:18308
Liquid
Extra Pure
1.0500g/mL
Authentic
1.4192 to 1.4212
180 to 220ppm MEHQ
139.0°C
13.0°C
Stabilized
98
C3H4O2
MFCD00004367
15, 121
NIXOWILDQLNWCW-UHFFFAOYSA-N
prop-2-enoic acid
6581
72.06
98%

Chemical Identifiers

79-10-7
72.063
NIXOWILDQLNWCW-UHFFFAOYSA-N
6581
prop-2-enoic acid
C3H4O2
MFCD00004367
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
CHEBI:18308
C=CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Very toxic to aquatic life.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : This product unavoidably polymerizes during storage

SDS Documents