Please login to your online account to display your discounted pricing

Acrylamide, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$389.40 - $389.40

Chemical Identifiers

Chemical Name or Material Acrylamide
CAS 79-06-1
Name Note Purified for Electrophoresis
Purity Grade Ultrapure Bioreagent
Molecular Formula CH2:CHCONH2
Formula Weight 71.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002199

 
avantor performance
408101
Poly Bottle 500g Each for $389.40
Description & Specifications

Specifications

Formula Weight 71.08
Chemical Name or Material Acrylamide
Molecular Formula CH2:CHCONH2
Name Note Purified for Electrophoresis
Quantity 500g
Purity Grade Ultrapure Bioreagent
Packaging Poly Bottle
Flash Point 138.00°C
Specific Gravity 1L = 1.12kg
CAS 79-06-1