Please login to your online account to display your discounted pricing

Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in, ACROS Organics™


Quantity 2.5L

Manufacturer: thermo fisher scientific  AC330200025

 View more versions of this product

Catalog No. AC330200025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes skin irritation. May cause genetic defects. May cause an allergic skin reaction. Causes serious eye irritation. May cause cancer. Harmful in contact with skin. Causes damage to organs through prolonged or repeated exposure. Harmful if swallowed. Suspected of damaging fertility. Harmful if inhaled.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Physical Form Liquid
Quantity 2.5L
Color Clear colorless
Purity Grade For biochemistry
Conductivity 3μS/cm Max.
Absorbance (1% in water)
Absorbance Conditions (1% in water)
CAS 110-26-9
Chemical Name or Material Acrylamide / N, N'-Methylenebisacrylamide 19:1
CAS 79-06-1
CAS 7732-18-5
CAS Max % 61.0
CAS 110-26-9
CAS Min % 59.0
Acrylic Acid 0.001% max.
MDL Number MFCD00080848
Health Hazard 1 Danger

Specifications

Chemical Name or Material Acrylamide / N, N'-Methylenebisacrylamide 19:1
CAS 110-26-9
CAS 7732-18-5
CAS Min % 59.0
CAS Max % 61.0
CAS 110-26-9
CAS 79-06-1
Purity Grade For biochemistry
MDL Number MFCD00080848
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Causes skin irritation. May cause genetic defects. May cause an allergic skin reaction. Causes serious eye irritation. May cause cancer. Harmful in contact with skin. Causes damage to organs through prolonged or repeated exposure. Harmful if swallowed. Suspected of damaging fertility. Harmful if inhaled.

SDS & Safety Documents