missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acrylamide/Bisacrylamide 29:1, powder

Premixed powder for preparing protein DNA gels

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
MDL Number MFCD00080848
UN Number UN2074
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6082417 View Documents Alfa Aesar™
J6082417
40g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6082422 View Documents Alfa Aesar™
J6082422
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6082428 View Documents Alfa Aesar™
J6082428
200g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

Solubility
It is soluble in water.

Notes
Keep away from light. Store at room tempeature.
Specifications

Specifications

Acrylamide/Bisacrylamide 29:1
MFCD00080848
UN2074