Please login to your online account to display your discounted pricing

Acrylamide/Bis Premix, Premix Ratio: 19:1, MP Biomedicals

$142.12 - $142.12

Chemical Identifiers

Chemical Name or Material Acrylamide/Bis Premix
Name Note Premix Ratio: 19:1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ICN800656

 
mp biomedicals
800656
Each for $142.12
Description & Specifications

Specifications

Quantity 200g
Name Note Premix Ratio: 19:1
Chemical Name or Material Acrylamide/Bis Premix