missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acrylamide-bis Premix 19:1, 5% Concentration, Acros Organics

Manufacturer:  Thermo Fisher Scientific 400270300

 View more versions of this product

Catalog No. AC400270300

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Acrylamide-bis premix 19:1
(1% in water) (1 cm cell)
0.05% max.
For biochemistry
95.0% acrylamide, 5.0% bisacrylamide
White
30g
110-26-9,79-06-1,110-26-9
25μS/cm (30% Max. in H2O)
(1% in water) (1 cm cell)
Fine Crystalline Powder
1.1
5 to 7 (10% aq. soln. at 20°C)

Chemical Identifiers

110-26-9,79-06-1,110-26-9
Acrylamide-bis premix 19:1
95.0% acrylamide, 5.0% bisacrylamide
For biochemistry

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Suspected of damaging fertility.

GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger