Please login to your online account to display your discounted pricing

Acrylamide-bis Premix 19:1, 5% Concentration, Acros Organics

Chemical Identifiers

Chemical Name or Material Acrylamide-bis premix 19:1
CAS 110-26-9
CAS 79-06-1
CAS 110-26-9
Purity Grade For biochemistry
Assay 95.0% acrylamide, 5.0% bisacrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC400271500

  SDS
thermo fisher scientific
400271500
150g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC400270300

  SDS
thermo fisher scientific
400270300
30g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Quantity 150g
Color White
Purity Grade For biochemistry
Assay 95.0% acrylamide, 5.0% bisacrylamide
Density 1.1
pH 5 to 7 (10% aq. soln. at 20°C)
Conductivity 25μS/cm (30% Max. in H2O)
CAS 110-26-9
CAS 110-26-9
Absorbance (1% in water) (1 cm cell)
Absorbance Conditions (1% in water) (1 cm cell)
CAS 79-06-1
Acrylic Acid 0.05% max.
Chemical Name or Material Acrylamide-bis premix 19:1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Suspected of damaging fertility.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Suspected of damaging fertility.