Please login to your online account to display your discounted pricing

Acrylamide, ≥99%, MP Biomedicals™


Quantity 1kg

Manufacturer: mp biomedicals  15025691

 View more versions of this product

Catalog No. ICN15025691

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 71.1
Color White
Quantity 1kg
Density 1.127g/cm3 at 25°C
Chemical Name or Material Acrylamide
Assay Percent Range ≥99%
Molecular Formula C3H5NO
CAS 79-06-1
Synonym 2-Propenamide

Specifications

Chemical Name or Material Acrylamide
CAS 79-06-1
Physical Form Crystalline Powder
Assay Percent Range ≥99%
Molecular Formula C3H5NO
Formula Weight 71.1
Synonym 2-Propenamide