Please login to your online account to display your discounted pricing

ACROS Organics™ Lysozyme, Egg White


Manufacturer: acros organics™  611930010

 View more versions of this product

Catalog No. AC611930010

  • / Each
 

Description & Specifications

Specifications

Quantity 1g
Source Egg white
Purity 1
Format Solid
Product Name Lysozyme