Please login to your online account to display your discounted pricing

ACROS Organics™ Glass Wool


Manufacturer: acros organics™  AC386060500

 View more versions of this product

Catalog No. AC386060500

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Filter aid
Quantity 50g
Boiling Point 1000°C
Product Type Glass Wool
Melting Point 680°C