Please login to your online account to display your discounted pricing

ACROS Organics™ Glass Wool


Manufacturer: acros organics™  AC386060010

 View more versions of this product

Catalog No. AC386060010

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Filter aid
Product Type Glass Wool
Boiling Point 1000°C
Melting Point 680°C
Quantity 1kg