Please login to your online account to display your discounted pricing

Alfa Aesar™ Acrolein diethyl acetal, 96%

Chemical Identifiers

Chemical Name or Material Acrolein diethyl acetal
CAS 3054-95-3
Molecular Formula CH2=CHCH(OCH2CH3)2
Formula Weight 130.19
MDL Number MFCD00009239
UN Number UN2374
Beilstein 1701567
Synonym 3,3-Diethoxy-1-propene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2482109

 
alfa aesar™
B2482109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2482118

 
alfa aesar™
B2482118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.19
Chemical Name or Material Acrolein diethyl acetal
Assay 96%
Molecular Formula CH2=CHCH(OCH2CH3)2
MDL Number MFCD00009239
CAS 3054-95-3
UN Number UN2374
Beilstein 1701567
Synonym 3,3-Diethoxy-1-propene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Boiling Point 124° to 125°C
Quantity 10g
Flash Point 13°C (55°F)
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 3,3-DIETHOXYPROPENE
RTECS Number AS1370000

Applications
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.