Please login to your online account to display your discounted pricing

Acrolein-Acrylonitrile Mixture (For EPA 603, 8030A/Appendix IX)


Quantity 1x1mL
Packaging Ampule

Manufacturer: ultra scientific  AMN6031

 View more versions of this product

Catalog No. US-AMN-603-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:
Acrolein
Acrylonitrile
(This solution is unstable; refrigerate and use immediately.)
Methods For EPA Methods 603, 8030A/Appendix IX
Chemical Name or Material Acrolein-Acrylonitrile Mixture
Packaging Ampule

Specifications

Chemical Name or Material Acrolein-Acrylonitrile Mixture