Please login to your online account to display your discounted pricing

Acrolein-Acrylonitrile Mixture (For EPA 603, 8030A/Appendix IX)

$55.20 - $55.20

Chemical Identifiers

Chemical Name or Material Acrolein-Acrylonitrile Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-AMN-603-1

 
ultra scientific
AMN6031
Ampule 1x1mL Each for $55.20
Description & Specifications

Specifications

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:
Acrolein
Acrylonitrile
(This solution is unstable; refrigerate and use immediately.)
Methods For EPA Methods 603, 8030A/Appendix IX
Chemical Name or Material Acrolein-Acrylonitrile Mixture
Packaging Ampule