Please login to your online account to display your discounted pricing

Acrolein, 96%, stab. with hydroquinone, Alfa Aesar

$149.18 - $149.18

Chemical Identifiers

Chemical Name or Material Acrolein
CAS 107-02-8
Physical Form Liquid
Assay Percent Range 96%
Molecular Formula H2C=CHCHO
Formula Weight 56.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36520AP

  SDS
alfa aesar
36520AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36520AE

  SDS
alfa aesar
36520AE
100mL Each for $149.18
Description & Specifications

Specifications

Formula Weight 56.06
Quantity 500mL
Density 0.84
Physical Form Liquid
Assay Percent Range 96%
Molecular Formula H2C=CHCHO
Chemical Name or Material Acrolein
CAS 107-02-8