missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acriflavine hydrochloride, Thermo Scientific Chemicals

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride

Supplier:  Thermo Scientific Chemicals 346840100

 View more versions of this product

Catalog No. AC346840100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acriflavine hydrochloride
260.0°C
7% max. (105°C, 1 hr.)
Glass bottle
MFCD00069039
22,I,65
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
MKLTXAHQKDVBLY-UHFFFAOYSA-N
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
53393742
69235-50-3
Brown-Red to Orange-Red
Authentic
C27H27Cl3N6
10 g
15,115
Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
541.91
Powder

Chemical Identifiers

69235-50-3
541.91
MKLTXAHQKDVBLY-UHFFFAOYSA-N
53393742
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
C27H27Cl3N6
MFCD00069039
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed s

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only