Learn More
Acriflavine hydrochloride, Thermo Scientific Chemicals
Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
$67.68 - $129.54
Chemical Identifiers
CAS | 69235-50-3 |
---|---|
Molecular Formula | C27H27Cl3N6 |
Molecular Weight (g/mol) | 541.91 |
MDL Number | MFCD00069039 |
InChI Key | MKLTXAHQKDVBLY-UHFFFAOYSA-N |
Synonym | unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride |
PubChem CID | 53393742 |
IUPAC Name | acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride |
SMILES | Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC346840100
|
Thermo Scientific Chemicals
346840100 |
10 g | Glass bottle |
Each for $67.68
|
|
||||
AC346840250
|
Thermo Scientific Chemicals
346840250 |
25 g | Glass bottle |
Each for $129.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69235-50-3 | |
541.91 | |
MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
53393742 | |
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
C27H27Cl3N6 | |
MFCD00069039 | |
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | |
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride |
Specifications
69235-50-3 | |
Brown-Red to Orange-Red | |
Authentic | |
C27H27Cl3N6 | |
10 g | |
15,115 | |
Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform | |
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 | |
541.91 | |
Powder |
260.0°C | |
7% max. (105°C, 1 hr.) | |
Glass bottle | |
MFCD00069039 | |
22,I,65 | |
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | |
MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride | |
53393742 | |
Acriflavine hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed s
GHS Signal Word: Danger
RUO – Research Use Only