missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acriflavine hydrochloride, ACROS Organics™

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride

Manufacturer:  Thermo Fisher Scientific 346840250

 View more versions of this product

Catalog No. AC346840250



Description

Description

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride

Specifications

Specifications

Acriflavine hydrochloride
7% max. (105°C, 1 hr.)
Brown-Red to Orange-Red
25g
C27H28Cl4N6
22,I,65
3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride, unii-1s73vw819c
VWXAJKFSYJFBIW-UHFFFAOYSA-N
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
53393742
Authentic
Glass bottle
260.0°C
69235-50-3
MFCD00069039
15,115
Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform
Cl.Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
578.36
Powder

Chemical Identifiers

69235-50-3
578.36
VWXAJKFSYJFBIW-UHFFFAOYSA-N
53393742
Cl.Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
C27H28Cl4N6
MFCD00069039
3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride, unii-1s73vw819c
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed s

GHS Signal Word: Danger

SDS Documents
Product Certifications