Learn More
Acridine-9-carboxylic acid hydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 400220050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acridine-9-carboxylic acid hydrate | |
96.0 | |
220.0°C | |
Authentic | |
Glass bottle | |
MFCD00149578 | |
22,V,3,413 | |
Solubility in water: soluble. Other solubilities: ethanol,pyridine,acetic acid,acetonitrile | |
OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12 | |
223.23 | |
241.25 | |
Crystalline Powder |
332927-03-4 | |
100.0 | |
Green-Yellow to Yellow | |
97% | |
C14H9NO2 | |
5 g | |
9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x | |
IYRYQBAAHMBIFT-UHFFFAOYSA-N | |
acridine-9-carboxylic acid;hydrate | |
16211687 | |
97% |
Chemical Identifiers
332927-03-4 | |
223.23 | |
IYRYQBAAHMBIFT-UHFFFAOYSA-N | |
16211687 |
C14H9NO2 | |
MFCD00149578 | |
9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x | |
OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
RUO – Research Use Only